Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010403.2 | NP_034533.1 | 1 - 2 | 158 | 75 | |
GenBank mRNA | AF104312.1 | - | 1 - 2 | 158 | 75 | |
AK149565.1 | - | 1 - 2 | 165 | 75 | ||
BC018587.1 | - | 1 - 2 | 136 | 75 | ||
BC119535.1 | - | 1 - 2 | 158 | 40056371 | 75 | |
BC119536.1 | - | 1 - 2 | 158 | 40056372 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |