Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042502.2 | NP_001035967.1 | 1 - 2 | 847 | 67 | |
NM_001287048.1 | NP_001273977.1 | 1 - 2 | 847 | 67 | ||
GenBank mRNA | AB006320.1 | - | 1 - 2 | 400 | 67 | |
AF048724.1 | - | 1 - 2 | 808 | 67 | ||
AF201091.1 | - | 1 - 2 | 174 | 67 | ||
AK035083.1 | - | 1 - 2 | 505 | 67 | ||
AM940439.1 | - | 1 - 2 | 100 | 67 | ||
BC075660.1 | - | 1 - 2 | 512 | 30606653 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |