Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282086.1 | NP_001269015.1 | 6 - 7 | 2056 | 81 | |
NM_013838.2 | NP_038866.2 | 7 - 8 | 2290 | 81 | ||
GenBank mRNA | AF057748.1 | - | 7 - 8 | 1859 | 81 | |
AK133864.1 | - | 6 - 7 | 1956 | 81 | ||
AK137447.1 | - | 6 - 7 | 1518 | 81 | ||
BC141131.1 | - | 7 - 8 | 2161 | 9055743 | 81 | |
U49069.1 | - | 7 - 8 | 2290 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |