Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163336.1 | NP_001156808.1 | 6 - 7 | 727 | 83 | |
NM_001163337.1 | NP_001156809.1 | 6 - 7 | 727 | 83 | ||
NM_016745.3 | NP_058025.2 | 6 - 7 | 727 | 83 | ||
GenBank mRNA | AK089487.1 | - | 6 - 7 | 727 | 83 | |
AK155904.1 | - | 6 - 7 | 728 | 83 | ||
BC017639.1 | - | 6 - 7 | 641 | 4924519 | 83 | |
BC026147.1 | - | 6 - 7 | 673 | 4191458 | 83 | |
U49393.1 | - | 6 - 7 | 588 | 83 | ||
U49394.1 | - | 6 - 7 | 588 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |