Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122733.1 | NP_001116205.1 | 7 - 8 | 1306 | 71 | |
NM_021099.3 | NP_066922.2 | 7 - 8 | 1302 | 71 | ||
GenBank mRNA | AK046795.1 | - | 7 - 8 | 1298 | 71 | |
AK047010.1 | - | 7 - 8 | 1302 | 71 | ||
AK145462.1 | - | 4 - 5 | 543 | 71 | ||
AY536430.1 | - | 7 - 8 | 1265 | 71 | ||
AY536431.1 | - | 7 - 8 | 1265 | 71 | ||
BC026713.1 | - | 7 - 8 | 1285 | 71 | ||
BC052457.1 | - | 7 - 8 | 1277 | 6830637 | 71 | |
BC075716.1 | - | 7 - 8 | 1291 | 3673641 | 71 | |
Y00864.1 | - | 7 - 8 | 1269 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Mouse Stem Cell Pluripotency 1700 Inventoried Mouse_Stem_Cell Probe spans exons |