Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021547.3 | NP_067522.1 | 12 - 13 | 1235 | 54 | |
GenBank mRNA | AK041538.1 | - | 12 - 13 | 1191 | 54 | |
AK150166.1 | - | 12 - 13 | 1179 | 54 | ||
AK150254.1 | - | 12 - 13 | 1179 | 54 | ||
AK155124.1 | - | 12 - 13 | 1174 | 54 | ||
AK160480.1 | - | 12 - 13 | 1155 | 54 | ||
BC003313.1 | - | 12 - 13 | 1152 | 3155179 | 54 | |
BC058727.1 | - | 13 - 14 | 1322 | 54 | ||
X82457.1 | - | 12 - 13 | 1219 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |