Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025494.3 | NP_079770.2 | 2 - 3 | 350 | 67 | |
GenBank mRNA | AB088407.1 | - | 2 - 3 | 297 | 67 | |
AK006323.1 | - | 2 - 3 | 301 | 67 | ||
AK075779.1 | - | 2 - 3 | 322 | 67 | ||
AK129007.1 | - | 2 - 3 | 310 | 67 | ||
AK151237.1 | - | 2 - 3 | 311 | 67 | ||
AK168914.1 | - | 2 - 3 | 301 | 67 | ||
BC010217.1 | - | 2 - 3 | 298 | 3498388 | 67 | |
U13839.1 | - | 2 - 3 | 293 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |