Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800016151.mm.1 |
NM_001113326.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016151.mm.1 |
NM_001113326.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016151.mm.1 |
NM_031195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016151.mm.1 |
NM_031195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016151.mm.1 |
TC0800002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016149.mm.1 |
NM_001113326.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016149.mm.1 |
NM_001113326.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016149.mm.1 |
NM_031195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016149.mm.1 |
NM_031195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016149.mm.1 |
TC0800002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016146.mm.1 |
NM_001113326.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016146.mm.1 |
NM_001113326.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016146.mm.1 |
NM_031195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016146.mm.1 |
NM_031195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016146.mm.1 |
TC0800002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016148.mm.1 |
NM_001113326.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016148.mm.1 |
NM_001113326.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016148.mm.1 |
NM_031195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016148.mm.1 |
NM_031195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016148.mm.1 |
TC0800002102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016144.mm.1 |
NM_001113326.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016144.mm.1 |
NM_001113326.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016144.mm.1 |
NM_031195.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016144.mm.1 |
NM_031195.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016144.mm.1 |
TC0800002102.mm.1 |
MTA TC |