Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080466.2 | NP_536714.2 | 4 - 5 | 1898 | 72 | |
GenBank mRNA | AF357241.1 | - | 4 - 5 | 1638 | 72 | |
AK137738.1 | - | 4 - 5 | 1899 | 72 | ||
BC106769.1 | - | 4 - 5 | 1640 | 40039722 | 72 | |
BC106770.1 | - | 4 - 5 | 1640 | 40039723 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |