Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cysltr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162412.1 | NP_001155884.1 | 1 - 2 | 75 | 96 | |
NM_133720.3 | NP_598481.2 | 1 - 2 | 75 | 96 | ||
GenBank mRNA | AF331658.1 | - | 1 - 2 | 34 | 96 | |
AK008997.1 | - | 1 - 2 | 76 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Mus musculus Inventoried Probe spans exons |