Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010833.2 | NP_034963.2 | 6 - 7 | 886 | 65 | |
GenBank mRNA | AK088336.1 | - | 6 - 7 | 834 | 65 | |
AK133819.1 | - | 6 - 7 | 890 | 65 | ||
AK145746.1 | - | 6 - 7 | 890 | 65 | ||
AK150869.1 | - | 6 - 7 | 889 | 65 | ||
AK153210.1 | - | 6 - 7 | 889 | 65 | ||
AK157701.1 | - | 6 - 7 | 841 | 65 | ||
AK216494.1 | - | 1 - 2 | 162 | 65 | ||
BC003474.1 | - | 6 - 7 | 877 | 65 | ||
BC047366.1 | - | 6 - 7 | 880 | 3711212 | 65 | |
CF584663.1 | - | 374 | 65 | |||
M86390.1 | - | 5 - 6 | 666 | 65 | ||
S47577.1 | - | 6 - 7 | 871 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |