Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011077.2 | NP_035207.1 | 4 - 5 | 962 | 75 | |
GenBank mRNA | AK158481.1 | - | 4 - 5 | 989 | 75 | |
BC137925.1 | - | 4 - 5 | 709 | 8860944 | 75 | |
BC137926.1 | - | 4 - 5 | 709 | 8860945 | 75 | |
EF194891.1 | - | 4 - 5 | 496 | 75 | ||
U49908.1 | - | 4 - 5 | 954 | 75 | ||
U73910.1 | - | 4 - 5 | 962 | 75 | ||
U75646.1 | - | 4 - 5 | 543 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |