Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011156.2 | NP_035286.1 | 10 - 11 | 1434 | 69 | |
GenBank mRNA | AB007631.1 | - | 10 - 11 | 1317 | 69 | |
AK053816.1 | - | 10 - 11 | 1445 | 69 | ||
AK076257.1 | - | 10 - 11 | 1433 | 69 | ||
AK133875.1 | - | 10 - 11 | 1433 | 69 | ||
AK141851.1 | - | 10 - 11 | 1443 | 69 | ||
AK161528.1 | - | 10 - 11 | 1433 | 69 | ||
AK163774.1 | - | 2 - 3 | 310 | 69 | ||
AK163787.1 | - | 10 - 11 | 1446 | 69 | ||
AK170570.1 | - | 10 - 11 | 1443 | 69 | ||
BC012869.1 | - | 10 - 11 | 1434 | 3967219 | 69 | |
BC050830.1 | - | 10 - 11 | 1427 | 6519113 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |