Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011703.4 | NP_035833.2 | 8 - 9 | 985 | 66 | |
GenBank mRNA | AF266282.1 | - | 8 - 9 | 867 | 66 | |
AK052465.1 | - | 8 - 9 | 986 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Mus musculus Probe spans exons |