Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011904.3 | NP_036034.1 | 8 - 9 | 1164 | 63 | |
GenBank mRNA | AF073526.1 | - | 8 - 9 | 1225 | 63 | |
BC132537.1 | - | 8 - 9 | 1164 | 40130814 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |