Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291152.1 | NP_001278081.1 | 7 - 8 | 745 | 70 | |
NM_013731.3 | NP_038759.1 | 8 - 9 | 863 | 70 | ||
GenBank mRNA | AF169033.1 | - | 8 - 9 | 798 | 70 | |
AK136559.1 | - | 8 - 9 | 815 | 70 | ||
BC026549.1 | - | 6 - 7 | 490 | 4973908 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |