Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827153 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827148 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827150 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827151 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827152 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827149 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2067181 |
AK013252.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2067181 |
BC021903.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2067181 |
NM_013919.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2067181 |
AK011148.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2067181 |
BP770216.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
450058 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
150766 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
8689 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P131938 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2067181 |
AK013252.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2067181 |
BC021903.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2067181 |
NM_013919.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2067181 |
AK011148.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2067181 |
BP770216.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827131 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827150 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827135 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827101 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827154 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827118 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827108 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827110 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827142 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827139 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827145 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827146 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827129 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827116 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827121 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827111 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827125 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827151 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827134 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827127 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827155 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827153 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827114 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827120 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827109 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827133 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827119 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827115 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827140 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827156 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827104 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827148 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827112 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827138 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P131938 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00250713 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P346747 |
AK013252.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P346747 |
AK207403.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P346747 |
BC021903.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P346747 |
NM_013919.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P346747 |
AF177759.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P116847 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P131938 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00289233 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00289290 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00289255 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00481671 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827153 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827148 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827152 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827149 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827151 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827054 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827101 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827108 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827103 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827129 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827118 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827135 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289240 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827153 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289276 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289242 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827142 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827133 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827125 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289269 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00853176 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827119 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827150 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827140 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827121 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00168121 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289233 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00481636 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827114 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01009891 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289236 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289255 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00481648 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827127 |
- |
- |