Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400020809.mm.1 |
TC0400002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020809.mm.1 |
NM_001190414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020809.mm.1 |
NM_001190414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020809.mm.1 |
NM_015828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020809.mm.1 |
NM_015828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020811.mm.1 |
TC0400002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020811.mm.1 |
NM_001190414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020811.mm.1 |
NM_001190414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020811.mm.1 |
NM_015828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020811.mm.1 |
NM_015828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020816.mm.1 |
TC0400002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020816.mm.1 |
NM_001190414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020816.mm.1 |
NM_001190414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020816.mm.1 |
NM_015828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020816.mm.1 |
NM_015828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020825.mm.1 |
TC0400002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020825.mm.1 |
NM_001190414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020825.mm.1 |
NM_001190414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020824.mm.1 |
TC0400002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020824.mm.1 |
NM_001190414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020824.mm.1 |
NM_001190414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020814.mm.1 |
TC0400002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020814.mm.1 |
NM_001190414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020814.mm.1 |
NM_001190414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020814.mm.1 |
NM_015828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020814.mm.1 |
NM_015828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020827.mm.1 |
TC0400002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020827.mm.1 |
NM_001190414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020827.mm.1 |
NM_001190414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020813.mm.1 |
TC0400002549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020813.mm.1 |
NM_001190414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020813.mm.1 |
NM_001190414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020813.mm.1 |
NM_015828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020813.mm.1 |
NM_015828.3 |
Refseq |