Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018889.3 | NP_061377.2 | 8 - 9 | 1132 | 96 | |
GenBank mRNA | AK153819.1 | - | 8 - 9 | 1049 | 96 | |
BC052658.1 | - | 8 - 9 | 1092 | 30048340 | 96 | |
D84436.1 | - | 8 - 9 | 1132 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |