Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019489.5 | NP_062362.1 | 3 - 4 | 186 | 83 | |
GenBank mRNA | AK008177.1 | - | 3 - 4 | 187 | 83 | |
BC045154.1 | - | 3 - 4 | 181 | 6468589 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |