Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019580.4 | NP_062526.1 | 4 - 5 | 726 | 106 | |
GenBank mRNA | AF212860.1 | - | 4 - 5 | 707 | 106 | |
AK005361.1 | - | 4 - 5 | 727 | 106 | ||
AK150807.1 | - | 4 - 5 | 748 | 106 | ||
BC003902.1 | - | 4 - 5 | 685 | 3601420 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |