Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252580.1 | NP_001239509.1 | 5 - 6 | 760 | 78 | |
NM_019729.3 | NP_062703.2 | 5 - 6 | 727 | 78 | ||
GenBank mRNA | AB045709.1 | - | 4 - 5 | 501 | 78 | |
AF057146.1 | - | 4 - 5 | 518 | 78 | ||
AK122195.1 | - | 5 - 6 | 638 | 78 | ||
AK129003.1 | - | 5 - 6 | 728 | 78 | ||
AK142881.1 | - | 2 - 3 | 225 | 78 | ||
AK163469.1 | - | 5 - 6 | 660 | 78 | ||
AK166540.1 | - | 5 - 6 | 728 | 78 | ||
BC038123.1 | - | 5 - 6 | 712 | 78 | ||
BC050947.1 | - | 5 - 6 | 692 | 6825981 | 78 | |
BC061465.1 | - | 5 - 6 | 692 | 6825981 | 78 | |
BC066126.1 | - | 5 - 6 | 690 | 30301665 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |