Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019810.4 | NP_062784.3 | 3 - 4 | 551 | 64 | |
GenBank mRNA | AF163846.1 | - | 3 - 4 | 313 | 64 | |
AF208031.1 | - | 3 - 4 | 551 | 64 | ||
AK033512.1 | - | 3 - 4 | 537 | 64 | ||
AK085602.1 | - | 3 - 4 | 342 | 64 | ||
BC003845.1 | - | 3 - 4 | 531 | 3497611 | 64 | |
U27269.1 | - | 1 - 2 | 121 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |