Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpgrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168515.1 | NP_001161987.1 | 9 - 10 | 1057 | 86 | |
NM_023879.3 | NP_076368.1 | 9 - 10 | 1057 | 86 | ||
GenBank mRNA | AK086397.1 | - | 6 - 7 | 751 | 86 | |
AY008297.1 | - | 9 - 10 | 1039 | 86 | ||
AY914560.1 | - | 9 - 10 | 1033 | 86 | ||
BC016092.1 | - | 9 - 10 | 1019 | 86 | ||
BC086692.1 | - | 9 - 10 | 1021 | 30682813 | 86 | |
HM067441.1 | - | 8 - 9 | 961 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |