Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22039671 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22039669 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22039670 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201904 |
AK004372.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201904 |
AK002279.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201904 |
NM_025316.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201904 |
AK004372.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201904 |
AK002279.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P201904 |
NM_025316.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103352 |
AK004372.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103352 |
AK013223.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103352 |
AK002279.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103352 |
NM_025316.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
627410 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
70120 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
750982 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
888849 |
AF016714.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
888849 |
NM_019680.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
961663 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
777819 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
927298 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
AK010507.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
AK013146.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
AK151058.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
AK151271.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
AK152361.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
AK152503.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
AK167385.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
AY562233.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
BC013052.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
NM_007874.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917268 |
U28168.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933230 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
476170 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
65697 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P116192 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P147569 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201904 |
AK004372.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201904 |
AK002279.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201904 |
NM_025316.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201904 |
AK004372.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201904 |
AK002279.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P201904 |
NM_025316.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103352 |
AK004372.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103352 |
AK013223.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103352 |
AK002279.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103352 |
NM_025316.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P201904 |
AK004372.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P201904 |
AK002279.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P201904 |
NM_025316.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P201904 |
AK004372.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P201904 |
AK002279.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P201904 |
NM_025316.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039670 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039688 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039706 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039682 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039671 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039677 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039704 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039679 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039697 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039678 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039669 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039683 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039687 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039696 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039674 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039702 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039693 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039699 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039691 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039690 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22039672 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00612859 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P147569 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04866189 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P116192 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P201904 |
AK004372.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201904 |
AK002279.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201904 |
NM_025316.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201904 |
AK004372.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201904 |
AK002279.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P201904 |
NM_025316.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P116180 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P147569 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P116192 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03191516 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02924713 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02924717 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03191503 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03191514 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22039669 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22039671 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22039670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22039722 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039711 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039715 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039677 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039670 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039674 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039672 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039671 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039692 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039675 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039699 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039704 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039710 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039683 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039697 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039696 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039718 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039679 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039707 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039668 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039709 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039693 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039695 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039669 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039703 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039682 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039706 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02350988 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039713 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039687 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039702 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039717 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22039700 |
- |
- |