Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029963.2 | NP_084239.1 | 1 - 2 | 61 | 67 | |
GenBank mRNA | AB049941.1 | - | 1 - 2 | 61 | 67 | |
AK018829.1 | - | 1 - 2 | 69 | 67 | ||
AK078198.1 | - | 1 - 2 | 69 | 67 | ||
AK083366.1 | - | 1 - 2 | 63 | 67 | ||
AK155101.1 | - | 1 - 2 | 77 | 67 | ||
BC021947.1 | - | 1 - 2 | 59 | 5098933 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |