Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138660.2 | NP_619601.2 | 5 - 6 | 655 | 77 | |
GenBank mRNA | AF526276.1 | - | 5 - 6 | 629 | 77 | |
AJ292072.1 | - | 869 | 77 | |||
AK139608.1 | - | 5 - 6 | 694 | 77 | ||
AK145886.1 | - | 5 - 6 | 429 | 77 | ||
BC034533.1 | - | 2 - 3 | 299 | 77 | ||
BC060672.1 | - | 5 - 6 | 483 | 77 | ||
BC141296.1 | - | 5 - 6 | 768 | 9055908 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |