Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144868.3 | NP_659117.2 | 24 - 25 | 4701 | 86 | |
GenBank mRNA | AF237953.1 | - | 22 - 23 | 2718 | 86 | |
AK220471.1 | - | 2 - 3 | 599 | 86 | ||
BC038056.1 | - | 19 - 20 | 2809 | 86 | ||
BC054751.1 | - | 24 - 25 | 3980 | 86 | ||
BC055289.1 | - | 25 - 26 | 4504 | 86 | ||
BC096538.1 | - | 24 - 25 | 4701 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |