Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025378.2 | NP_001020549.1 | 6 - 7 | 568 | 113 | |
NM_001271526.1 | NP_001258455.1 | 9 - 10 | 1093 | 113 | ||
NM_008765.3 | NP_032791.1 | 9 - 10 | 1099 | 113 | ||
GenBank mRNA | AB190256.1 | - | 6 - 7 | 558 | 113 | |
AK048408.1 | - | 9 - 10 | 918 | 113 | ||
AK051747.1 | - | 9 - 10 | 928 | 113 | ||
AK133604.1 | - | 9 - 10 | 919 | 113 | ||
AK146281.1 | - | 9 - 10 | 918 | 113 | ||
AK146859.1 | - | 9 - 10 | 912 | 113 | ||
BC015257.1 | - | 9 - 10 | 909 | 4234871 | 113 | |
U27457.1 | - | 9 - 10 | 918 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |