Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009619.4 | NP_033749.2 | 6 - 7 | 600 | 116 | |
GenBank mRNA | BC060975.1 | - | 6 - 7 | 563 | 116 | |
BC129920.1 | - | 6 - 7 | 566 | 116 | ||
X64227.1 | - | 6 - 7 | 610 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |