Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301211.1 | NP_001288140.1 | 4 - 5 | 856 | 75 | |
NM_009677.6 | NP_033807.2 | 4 - 5 | 856 | 75 | ||
GenBank mRNA | AK034198.1 | - | 1 - 2 | 85 | 75 | |
AK036387.1 | - | 4 - 5 | 799 | 75 | ||
AK145821.1 | - | 4 - 5 | 778 | 75 | ||
BC006691.1 | - | 4 - 5 | 74 | |||
BC052703.1 | - | 4 - 5 | 729 | 5698075 | 75 | |
BC054535.1 | - | 4 - 5 | 729 | 5698075 | 75 | |
X54424.1 | - | 4 - 5 | 778 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |