Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011698.1 | NP_035828.1 | 2 - 3 | 159 | 71 | |
GenBank mRNA | AF087694.1 | - | 2 - 3 | 166 | 71 | |
AF173082.1 | - | 2 - 3 | 159 | 71 | ||
AK019299.1 | - | 2 - 3 | 164 | 71 | ||
BC031780.1 | - | 2 - 3 | 178 | 4164883 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |