Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vill | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164567.1 | NP_001158039.1 | 15 - 16 | 1868 | 119 | |
NM_011700.2 | NP_035830.2 | 15 - 16 | 1868 | 119 | ||
GenBank mRNA | AJ344341.1 | - | 15 - 16 | 1856 | 119 | |
AK028229.1 | - | 15 - 16 | 1869 | 119 | ||
AK166652.1 | - | 4 - 5 | 621 | 119 | ||
BC021808.1 | - | 15 - 16 | 1844 | 3991439 | 119 | |
BC022664.1 | - | 6 - 7 | 797 | 4223317 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |