Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc93b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161428.1 | NP_001154900.1 | 4 - 5 | 627 | 78 | |
NM_019449.2 | NP_062322.2 | 4 - 5 | 627 | 78 | ||
GenBank mRNA | AK030488.1 | - | 4 - 5 | 588 | 78 | |
AK155556.1 | - | 4 - 5 | 585 | 78 | ||
AK162535.1 | - | 4 - 5 | 580 | 78 | ||
AK165060.1 | - | 4 - 5 | 587 | 78 | ||
BC018388.1 | - | 4 - 5 | 594 | 4209296 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |