Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023214.7 | NP_075703.1 | 2 - 3 | 376 | 64 | |
GenBank mRNA | AF233322.1 | - | 2 - 3 | 238 | 64 | |
AF529196.1 | - | 2 - 3 | 184 | 64 | ||
AK010008.1 | - | 2 - 3 | 376 | 64 | ||
AK075802.1 | - | 2 - 3 | 349 | 64 | ||
BC017136.1 | - | 2 - 3 | 326 | 4911068 | 64 | |
BC050193.1 | - | 2 - 3 | 275 | 30019802 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |