Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033136.1 | 1 - 2 | 257 | 86 | ||
GenBank mRNA | AF310624.1 | - | 1 - 2 | 198 | 86 | |
AF312938.1 | - | 1 - 2 | 153 | 86 | ||
AF375475.1 | - | 1 - 2 | 182 | 86 | ||
AK131939.1 | - | 1 - 2 | 198 | 86 | ||
AK146479.1 | - | 1 - 2 | 199 | 86 | ||
AK159624.1 | - | 1 - 2 | 202 | 86 | ||
BC092068.1 | - | 1 - 2 | 182 | 4210931 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |