Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031381.2 | NP_113558.1 | 1 - 2 | 62 | 69 | |
GenBank mRNA | AF285576.1 | - | 1 - 2 | 62 | 69 | |
BC116863.1 | - | 1 - 2 | 38 | 40126182 | 69 | |
BC119125.1 | - | 1 - 2 | 38 | 8733874 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |