Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002238.2 | NP_001002238.1 | 13 - 14 | 1727 | 78 | |
NM_001002240.2 | NP_001002240.1 | 13 - 14 | 1700 | 78 | ||
NM_001002241.2 | NP_001002241.1 | 12 - 13 | 2000 | 78 | ||
NM_031387.3 | NP_113564.2 | 13 - 14 | 1860 | 78 | ||
GenBank mRNA | AB067571.1 | - | 12 - 13 | 1681 | 78 | |
AB183526.1 | - | 13 - 14 | 1727 | 78 | ||
AB183527.1 | - | 13 - 14 | 1700 | 78 | ||
AB183528.1 | - | 12 - 13 | 2000 | 78 | ||
AB183529.1 | - | 12 - 13 | 1564 | 78 | ||
AF285591.1 | - | 9 - 10 | 1054 | 78 | ||
AK029806.1 | - | 13 - 14 | 1861 | 78 | ||
BC129954.1 | - | 8 - 9 | 986 | 40081321 | 78 | |
BC129955.1 | - | 8 - 9 | 986 | 40081323 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |