Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291010.1 | NP_001277939.1 | 2 - 3 | 317 | 79 | |
NM_033042.4 | NP_149031.2 | 1 - 2 | 378 | 79 | ||
GenBank mRNA | AF329969.1 | - | 2 - 3 | 321 | 79 | |
BC017526.1 | - | 1 - 2 | 378 | 4912933 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |