Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271347.1 | NP_001258276.1 | 7 - 8 | 1250 | 89 | |
NM_001271348.1 | NP_001258277.1 | 7 - 8 | 1211 | 89 | ||
NM_001271349.1 | NP_001258278.1 | 6 - 7 | 1148 | 89 | ||
NM_134015.3 | NP_598776.1 | 8 - 9 | 1313 | 89 | ||
GenBank mRNA | AB093260.1 | - | 6 - 7 | 955 | 89 | |
AK041532.1 | - | 7 - 8 | 1043 | 89 | ||
AK149139.1 | - | 7 - 8 | 1033 | 89 | ||
AK152181.1 | - | 7 - 8 | 973 | 89 | ||
AK160086.1 | - | 6 - 7 | 949 | 89 | ||
AK166226.1 | - | 7 - 8 | 908 | 89 | ||
AK168667.1 | - | 6 - 7 | 965 | 89 | ||
AK218523.1 | - | 1 - 2 | 121 | 89 | ||
AY038079.1 | - | 8 - 9 | 977 | 89 | ||
BC008552.1 | - | 1 - 2 | 46 | 89 | ||
BC034261.1 | - | 8 - 9 | 993 | 4237375 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |