Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144800.2 | NP_659049.2 | 12 - 13 | 1746 | 81 | |
GenBank mRNA | AK030533.1 | - | 10 - 11 | 1140 | 81 | |
AY214918.1 | - | 12 - 13 | 1247 | 81 | ||
BC016555.1 | - | 1 - 2 | 61 | 81 | ||
BC023682.1 | - | 6 - 7 | 852 | 81 | ||
BC024131.1 | - | 12 - 13 | 1711 | 5101888 | 81 | |
BC042632.1 | - | 1 - 2 | 65 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |