Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145477.2 | NP_663452.1 | 5 - 6 | 773 | 77 | |
GenBank mRNA | BC021379.1 | - | 5 - 6 | 746 | 5044319 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |