Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152944.1 | NP_694423.1 | 3 - 4 | 851 | 62 | |
GenBank mRNA | AF507967.1 | - | 3 - 4 | 836 | 62 | |
AY124569.1 | - | 3 - 4 | 851 | 62 | ||
BC106860.2 | - | 3 - 4 | 851 | 40047272 | 62 | |
BC106861.2 | - | 3 - 4 | 851 | 40047279 | 62 | |
BC111522.1 | - | 2 - 3 | 1658 | 40047286 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |