Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153056.2 | NP_694696.2 | 1 - 2 | 123 | 97 | |
GenBank mRNA | AK051725.1 | - | 1 - 2 | 113 | 97 | |
AK135484.1 | - | 1 - 2 | 120 | 97 | ||
AY251540.1 | - | 1 - 2 | 99 | 97 | ||
BC026403.1 | - | 1 - 2 | 88 | 4209498 | 97 | |
BC026637.1 | - | 1 - 2 | 91 | 97 | ||
BC026650.1 | - | 1 - 2 | 91 | 4987746 | 97 | |
BC040759.1 | - | 1 - 2 | 88 | 4209498 | 97 | |
BY125424.1 | - | 1 - 2 | 135 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |