Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199146.1 | NP_001186075.1 | 4 - 5 | 688 | 58 | |
NM_001199147.1 | NP_001186076.1 | 4 - 5 | 757 | 58 | ||
NM_153410.5 | NP_700459.2 | 4 - 5 | 688 | 58 | ||
GenBank mRNA | AK048691.1 | - | 4 - 5 | 682 | 58 | |
AK082985.1 | - | 4 - 5 | 631 | 58 | ||
AK084631.1 | - | 4 - 5 | 630 | 58 | ||
BC026486.1 | - | 4 - 5 | 748 | 58 | ||
BC027004.1 | - | 2 - 3 | 195 | 58 | ||
BC071197.1 | - | 4 - 5 | 754 | 6465025 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |