Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153415.3 | NP_700464.2 | 3 - 4 | 921 | 59 | |
GenBank mRNA | AF246235.1 | - | 3 - 4 | 525 | 59 | |
AK043573.1 | - | 3 - 4 | 622 | 59 | ||
AK043713.1 | - | 3 - 4 | 463 | 59 | ||
AK050915.1 | - | 3 - 4 | 490 | 59 | ||
AK076813.1 | - | 3 - 4 | 923 | 59 | ||
AK085703.1 | - | 3 - 4 | 990 | 59 | ||
AY090481.1 | - | 3 - 4 | 921 | 59 | ||
AY090482.1 | - | 3 - 4 | 521 | 59 | ||
BC052045.1 | - | 3 - 4 | 493 | 6402328 | 59 | |
BC062965.1 | - | 3 - 4 | 493 | 59 | ||
BC079589.1 | - | 3 - 4 | 491 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |