Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800012130.mm.1 |
NM_153775.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012130.mm.1 |
NM_153775.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012130.mm.1 |
TC0800001490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012118.mm.1 |
NM_153775.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012118.mm.1 |
NM_153775.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012118.mm.1 |
TC0800001490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012125.mm.1 |
NM_153775.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012125.mm.1 |
NM_153775.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012125.mm.1 |
TC0800001490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012123.mm.1 |
NM_153775.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012123.mm.1 |
NM_153775.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012123.mm.1 |
TC0800001490.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012128.mm.1 |
NM_153775.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012128.mm.1 |
NM_153775.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012128.mm.1 |
TC0800001490.mm.1 |
MTA TC |