Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011082.3 | NP_035212.2 | 7 - 8 | 2079 | 67 | |
GenBank mRNA | AK128944.1 | - | 7 - 8 | 2079 | 67 | |
AK142656.1 | - | 7 - 8 | 2081 | 67 | ||
BC013556.1 | - | 7 - 8 | 2012 | 3601095 | 67 | |
U06431.1 | - | 7 - 8 | 1983 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |