Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011159.2 | NP_035289.2 | 27 - 28 | 3281 | 105 | |
GenBank mRNA | AB007544.1 | - | 27 - 28 | 3258 | 105 | |
AB011543.1 | - | 27 - 28 | 3270 | 105 | ||
D87521.1 | - | 27 - 28 | 3258 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |