Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054387 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054390 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054389 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054388 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054386 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P190647 |
AK014516.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P190647 |
AK014516.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
903658 |
BC111822.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
903658 |
NM_053219.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
558517 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
175419 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P176486 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P190647 |
AK014516.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P190647 |
AK014516.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P190647 |
AK014516.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P190647 |
AK014516.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315530 |
AK086190.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315530 |
AK138742.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315530 |
BC051970.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315530 |
U32394.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315530 |
NM_016662.4 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054408 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054368 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054401 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054394 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054379 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054377 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054404 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054409 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054389 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054355 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054387 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054406 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054405 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054354 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054352 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054371 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054396 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054356 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054361 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054376 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054392 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054385 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054365 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054369 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054360 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054384 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054400 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054398 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054374 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054402 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054390 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054381 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29054380 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07882526 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P176486 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P190647 |
AK014516.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P190647 |
AK014516.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P315530 |
AK086190.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P315530 |
AK138742.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P315530 |
BC051970.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P315530 |
U32394.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P315530 |
NM_016662.4 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P118708 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P176486 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10610123 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10821703 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10610120 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10821707 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10610127 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10610122 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054389 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054387 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054388 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054386 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29054390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054304 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054334 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054342 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054302 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054329 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054306 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054319 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054324 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054301 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054322 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054331 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054392 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054316 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054328 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054344 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054340 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29054406 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054380 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054381 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10777838 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054366 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11057954 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10777836 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054360 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054384 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054417 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054355 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054408 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054361 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11109886 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054395 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054387 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054352 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054396 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10531359 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10723420 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054356 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11054964 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054376 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11109885 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10821706 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054390 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054392 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054389 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054391 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054377 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054413 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054400 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054409 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11057943 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054404 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10438504 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10600441 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10900329 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054362 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054416 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054379 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29054371 |
- |
- |